Learn More
American Research Products Inc Mouse Kidney injury molecule 1
Supplier:  American Research Products Inc CSBE08809M
Encompass
Mouse Kidney injury molecule 1 Kim-1 ELISA Kit from CUSABIO Detection Range 0 9 ng/mL-60 ng/mL Sensitivity 0 225 ng/mL Sample serum plasma tissue homogenates urine Method Sandwich
Catalog No. 50-003-26724