Learn More
Crystal Chem Inc Mouse Kidney Injury Molecule-1 (KIM-1) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80652
Encompass
The Mouse KIM-1 ELISA is a highly sensitive assay that is used to quantify levels of mouse kim-1 in plasma, serum, and urine.Highlights: Fast test results in just 3 hoursReliable and accurate: CV's < 20%. Dynamic range of 0 to 5 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7782