Learn More
BIOHIPPO Mouse Kim1 Microsample
SDP
Catalog No. 5000315770
Encompass
Mouse Kim1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Kim1 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluidstissue homogenates It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 135 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Kim1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Kim1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Kim1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15770 Supplier BIOHIPPO Supplier No. BHE15212721
Mouse Kim1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Kim1 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluidstissue homogenates It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 135 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Kim1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Kim1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Kim1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color