Learn More
R&D Systems™ Mouse KIR3DL1 (AAN05414.1) VersaClone cDNA

Description
Specifications
Specifications
| Gene | KIR3DL1 |
| Accession Number | AAN05414.1 |
| Concentration | 10 μg at 0.2 μg/ μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1312 bp |
| Gene ID (Entrez) | 3811 |
| Gene Alias | AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3 DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3 DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, Nkat3, NKAT-3, NKAT3KIR, NKB1, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.