Learn More
Sino Biological Mouse KIRREL1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50193T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse KIRREL1 / NEPH1 (Catalog#50193-M08H; NP_570937.2; Met1-Leu525). KIRREL1 / NEPH1 specific IgG was purified by Mouse KIRREL1 / NEPH1 affinity chromatography.
Catalog No. 50-237-0915