Learn More
Sino Biological Mouse KIT MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50530R017100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse KIT / c-KIT / CD117 (rM KIT / c-KIT / CD117; Catalog#50530-M08H; NP_001116205.1; Met1-Thr523).
Catalog No. 50-237-2469