Learn More
Sino Biological Anti-c-Kit Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50530T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse KIT / c-KIT / CD117 (Catalog#50530-M08H; NP_001116205.1; Met1-Thr523). KIT / c-KIT / CD117 specific IgG was purified by Mouse KIT / c-KIT / CD117 affinity chromatography.
Catalog No. 50-240-8848