Learn More
Sino Biological Anti-Kallikrein 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50915T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse KLK1 / Kallikrein 1 (rM KLK1 / Kallikrein 1; Catalog#50915-M08H; P15947; Met1-Asp261). KLK1 / Kallikrein 1 specific IgG was purified by Mouse KLK1 / Kallikrein 1 affinity chromatography.
Catalog No. 50-255-3051