Learn More
Sino Biological Mouse KLK15 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 55382M08H100
Encompass_Preferred
A DNA sequence encoding the Mouse KLK15 (NP_777354.1) (Gln15-Tyr254) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2537