Learn More
Sino Biological Anti-Kallikrein 6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50255RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse KLK6 / Kallikrein 6 / Neurosin (NP_035307.2; Met 8-Leu 253). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3326