Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse KLRA2 Synthetic Peptide

Catalog No. PEP0803
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0803 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0803 Supplier Invitrogen™ Supplier No. PEP0803
Only null left
Add to Cart
Add to Cart

Synthetic Peptide

KLRA2 Synthetic Peptide, PEP-0803, from Invitrogen.

KLRA2 is a gene that encodes a protein called Killer Cell Lectin-like Receptor A2. This protein is expressed on the surface of natural killer cells and is involved in recognizing and eliminating virally infected or cancerous cells. KLRA2 is located on chromosome 12 and is part of a larger family of genes that encode for similar receptors. The KLRA2 protein interacts with a variety of ligands, including certain viruses and tumor cells. Mutations or variations in the KLRA2 gene have been associated with autoimmune disorders and cancer. KLRA2 is an important gene that plays a key role in the immune system's ability to recognize and eliminate abnormal or infected cells.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 16633
Species Mouse
Purification Method purified
Accession Number Q60660
Common Name Mouse KLRA2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol Klra2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.