Learn More
Sino Biological Mouse KLRB1F Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50815M01H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse KLRB1F (Q8VD98) (Gln 67-Val 217) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-0481