Learn More
Sino Biological Anti-NKG2A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50834RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse NKG2A / NKG2 / CD159A / KLRC1 (rM NKG2A / NKG2 / CD159A / KLRC1; Catalog#50834-M07H; AAD24969.1; Ala94-Ile244). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3062