Learn More
Sino Biological Recombinant Mouse Kininogen 1 Protein (His Tag)
Supplier:  Sino Biological 50474M08H50
Encompass_Preferred
A DNA sequence encoding the mouse KNG1 isoform 3 (NP_001095882.1) (Glu 21-Ser 480) was expressed, with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-224-7166