Learn More
Sino Biological Anti-Kininogen 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50474T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse KNG1/Kininogen 1 (rM KNG1/Kininogen 1; Catalog#50474-M08H; NP_001095882.1; Glu21-Ser480). KNG1/Kininogen 1 specific IgG was purified by Mouse KNG1/Kininogen 1 affinity chromatography.
Catalog No. 50-241-1821