Learn More
Sino Biological Anti-Kininogen 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50474RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse KNG1 (rM KNG1; Catalog#50474-M08H; NP_001095882.1; Glu 21-Ser 480). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1772