Learn More
BIOHIPPO Mouse KRT18 Microsample
SDP
Catalog No. 5000314826
Encompass
Mouse KRT18 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of KRT18 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse KRT18 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse KRT18 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse KRT18 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14826 Supplier BIOHIPPO Supplier No. BHE15211537
Mouse KRT18 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of KRT18 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse KRT18 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse KRT18 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse KRT18 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color