Learn More
Sino Biological Recombinant Anti-kynureninase/KYNU Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50310R004100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse KYNU (rM KYNU; Catalog#50310-M08B; Q9CXF0; Met1-Ser464).
Catalog No. 50-241-1920