Learn More
Sino Biological Mouse L-selectin/CD62L Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785742
Encompass_Preferred
The Mouse SELL / CD62L / L-Selectin ELISA Pair Set is for the quantitative determinationof Mouse SELL / CD62L / L-Selectin.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for SELL / CD62L / L-Selectin coated on a 96-well plate. Standards and samples are added to the wells, and any SELL / CD62L / L-Selectin present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-SELL / CD62L / L-Selectin polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of SELL / CD62L / L-Selectin present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwel
Catalog No. 50-007-85742 Supplier Sino Biological Supplier No. SEK500455
The Mouse SELL / CD62L / L-Selectin ELISA Pair Set is for the quantitative determinationof Mouse SELL / CD62L / L-Selectin.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for SELL / CD62L / L-Selectin coated on a 96-well plate. Standards and samples are added to the wells, and any SELL / CD62L / L-Selectin present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-SELL / CD62L / L-Selectin polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of SELL / CD62L / L-Selectin present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwel