Learn More
Sino Biological Mouse L1CAM Protein 1MG (Sino Biological)
Supplier:  Sino Biological 50835M02H1
Encompass_Preferred
A DNA sequence encoding the Mouse L1CAM (P11627) (Met1-Glu1123) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1575