Learn More
Raybiotech Inc L-Series Mouse Antibody Array 2 Glass Slide Kit.
Catalog No. 501948682
Encompass_Preferred
Detects 500 Mouse Proteins. Suitable for serum, plasma and cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAM-BLG-2-4
Catalog No. 50-194-8682 Supplier Raybiotech Inc Supplier No. AAMBLG24
Detects 500 Mouse Proteins. Suitable for serum, plasma and cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAM-BLG-2-4