Learn More
Raybiotech Inc L-Series Mouse Antibody Array 308 Glass Slide Kit.
Catalog No. 501948681
Encompass_Preferred
Detects 308 Mouse Proteins. Suitable for serum, plasma and cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAM-BLG-1-8
Catalog No. 50-194-8681 Supplier Raybiotech Inc Supplier No. AAMBLG18
Detects 308 Mouse Proteins. Suitable for serum, plasma and cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAM-BLG-1-8