Learn More
ACROBiosystems Recombinant Protein;Mouse LAG-3 / CD223 Protein;HEK293;100UG;LA3,LAG3,CD223,LAG-3,FDC
Supplier:  ACROBiosystems LA3M52H5100UG
Encompass_Preferred
Recombinant Protein;Mouse LAG-3 / CD223 Protein;100UG;Mouse LAG-3, His Tag (LA3-M52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 442 (Accession # Q61790).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.1 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-108-9647