Learn More
Bon Opus Biosciences Recombinant Mouse LAG3 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP101750UG
Encompass_Preferred
Recombinant Mouse LAG3 (C-hFc) is produced by Human Cells. The target gene encoding Ser23-LEU442 is expressed with a C-hFc tag. Accession# Q61790. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1017
Catalog No. 50-217-2619