Learn More
Sino Biological Recombinant Mouse LAG-3 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 53069M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse LAG3 (NP_032505.1)(Met1-Leu442) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2965