Learn More
Sino Biological Anti-LAIR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51030RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LAIR1 (Catalog#51030-M08H; Q8BG84-1; Gln22-Ser133). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3024