Learn More
Sino Biological Mouse LAIR1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 51030T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LAIR1 (Catalog#51030-M08H; Q8BG84-1; Gln22-Ser133). LAIR1 specific IgG was purified by Mouse LAIR1 affinity chromatography.
Catalog No. 50-237-3342