Learn More
Biomatik Corporation Mouse Laminin Alpha 5 (LAMa5) ELISA Kit
Supplier:  Biomatik Corporation EKU08501
Encompass_Preferred
Mouse Laminin Alpha 5 (LAMa5) ELISA Kit for detecting LAMa5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61001. Gene ID: 16776. Target Synonyms: LAM-A5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8659