Learn More
Biomatik Corporation Mouse Laminin (LN) CLIA Kit
Supplier:  Biomatik Corporation EKU08811
Encompass_Preferred
Mouse Laminin (LN) CLIA Kit for detecting LN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P19137; P02469; P02468. Gene ID: 16772; 16777. Target Synonyms: LAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7409