Learn More
Biomatik Corporation Mouse Laminin (LN) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12116
Encompass_Preferred
Mouse Laminin (LN) Mini sample ELISA Kit for detecting LN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19137; P02469; P02468. Gene ID: 16772; 16777. Target Synonyms: LAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5355