Learn More
Sino Biological Recombinant Anti-LAMP2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50791R141100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LAMP2/CD107b (rM LAMP2/CD107b; Catalog#50791-M08H; P17047-1; Leu26-Asn379).
Catalog No. 50-255-3094