Learn More
Sino Biological Mouse LAMP2 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 50791R141P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LAMP2/CD107b (rM LAMP2/CD107b; Catalog#50791-M08H; P17047-1; Leu26-Asn379) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-2812