Learn More
Sino Biological Anti-LAMP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50791T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LAMP2 / CD107b (Catalog#50791-M08H; P17047-1; Leu26-Asn379). LAMP2 / CD107b specific IgG was purified by Mouse LAMP2 / CD107b affinity chromatography.
Catalog No. 50-255-3093