Learn More
Biomatik Corporation Mouse Large Multifunctional Peptidase 7 (LMP7) ELISA Kit
Supplier:  Biomatik Corporation EKU11697
Encompass_Preferred
Mouse Large Multifunctional Peptidase 7 (LMP7) ELISA Kit for detecting LMP7 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28063. Gene ID: 16913. Target Synonyms: PSMB8; RING10; PSMB5i, Beta5i; Proteasome (Prosome,Macropain) Subunit,Beta Type 8; Multicatalytic endopeptidase complex subunit C13; Really interesting new gene 10 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7365