Learn More
ACROBiosystems ACROBiosystems Mouse Latent TGF-beta 1 / TGFB1 Protein, His Tag
Catalog No. 502106543
Encompass_Preferred
Recombinant Protein;1MG;Mouse Latent TGFB1, His Tag (TG1-M5245) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P04202-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 12 kDa and 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Latent TGFB1, His Tag (Cat. No. TG1-M5245) at 2 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB8 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT8-H82W5) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Mouse Latent TGFB1, His Tag (Cat. No. TG1-M5245) at 2 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 1-20 ng/mL (Routinely tested).
Catalog No. 50-210-6543 Supplier ACROBiosystems Supplier No. TG1M52451MG
Recombinant Protein;1MG;Mouse Latent TGFB1, His Tag (TG1-M5245) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P04202-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 12 kDa and 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Latent TGFB1, His Tag (Cat. No. TG1-M5245) at 2 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB8 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT8-H82W5) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Mouse Latent TGFB1, His Tag (Cat. No. TG1-M5245) at 2 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 1-20 ng/mL (Routinely tested).