Learn More
Biomatik Corporation Mouse Latexin (LXN) ELISA Kit
Supplier:  Biomatik Corporation EKU11036
Encompass_Preferred
Mouse Latexin (LXN) ELISA Kit for detecting LXN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70202. Gene ID: 17035. Target Synonyms: ECI; TCI; MUM; Endogenous carboxypeptidase inhibitor; Tissue carboxypeptidase inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8242