Learn More
Sino Biological Anti-LDHA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51207T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LDH-A / LDHA (rM LDH-A / LDHA; Catalog#51207-M07E; NP_034829.1; Met1-Phe332). LDH-A / LDHA specific IgG was purified by Mouse LDH-A / LDHA affinity chromatography.
Catalog No. 50-255-2969