Learn More
Sino Biological Recombinant Anti-LDLR/LDL Receptor Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50305R101100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LDLR (rM LDLR; Catalog#50305-M08H; NP_034830.2; Met1-Arg790).
Catalog No. 50-241-2454