Learn More
Sino Biological Anti-LDLR/LDL Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50305RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LDLR / LDL Receptor (rM LDLR; Catalog#50305-M08H; NP_034830.2; Met 1-Arg 790). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1728