Learn More
Sino Biological Recombinant Mouse LDLR/LDL Receptor Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50305M08HB20
Encompass_Preferred
A DNA sequence encoding the mouse LDLR (NP_034830.2) precursor (Met1-Arg790) with substitution of Val 23 and Gly 27 by Ala 23 and Cys 27 respectively was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-3313