Learn More
BIOHIPPO Mouse LECT1 Microsample
SDP
Catalog No. 5000314919
Encompass
Mouse LECT1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LECT1 in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LECT1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LECT1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LECT1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14919 Supplier BIOHIPPO Supplier No. BHE15211198
Mouse LECT1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LECT1 in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LECT1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LECT1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LECT1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color