Learn More
Biomatik Corporation Mouse Left/Right Determination Factor 2 (LEFTY2) ELISA Kit
Supplier:  Biomatik Corporation EKU05584
Encompass_Preferred
Mouse Left/Right Determination Factor 2 (LEFTY2) ELISA Kit for detecting LEFTY2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P57785. Gene ID: 320202. Target Synonyms: TGFB4; EBAF; LEFTA; LEFTYA; Endometrial Bleeding Associated Factor; Left-right determination factor A; Transforming growth factor beta-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6996