Learn More
Sino Biological Mouse LEP Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50442M38H50
Encompass_Preferred
A DNA sequence encoding the mouse Leptin (P41160) (Met1-Cys167) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-237-3586