Learn More
Biomatik Corporation Mouse Leptin (LEP) CLIA Kit
Supplier:  Biomatik Corporation EKU08814
Encompass_Preferred
Mouse Leptin (LEP) CLIA Kit for detecting LEP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 41.2-30,000pg/ml. Sensitivity: 13.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P41160. Gene ID: 16846. Target Synonyms: OB; OBS; Obesity Homolog; Obesity Factor; Obese Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8623