Learn More
Bioworld MOUSE LEPTIN TRIPLE ANTAGON
Supplier:  Bioworld 220606011
Encompass_Preferred
MOUSE LEPTIN TRIPLE ANTAGONIST (RECOMBINANT): Recombinant mouse leptin (mLEP) antagonist (triple mutant) Source: E.Coli Background: Leptin inhibits food intake and stimulates energy expenditure. Leptin also has thermogenic actions an...
Catalog No. 50-253-701