Learn More
Biomatik Corporation Mouse Leucine Rich Repeat Containing G Protein Coupled Receptor 5 (LGR5) ELISA Kit
Supplier:  Biomatik Corporation EKU12347
Encompass_Preferred
Mouse Leucine Rich Repeat Containing G Protein Coupled Receptor 5 (LGR5) ELISA Kit for detecting LGR5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z1P4. Gene ID: 14160. Target Synonyms: FEX; GPR49; GPR67; HG38; G Protein-Coupled Receptor 49. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5840