Learn More
Sino Biological Anti-Galectin-1/LGALS1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50100RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Galectin-1 / LGALS1 (rM Galectin-1 / LGALS1; Catalog#50100-MNAE; P16045; Met1-Glu135). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3462