Learn More
BIOHIPPO Mouse LGALS3BP Microsample
SDP
Catalog No. 5000315046
Encompass
Mouse LGALS3BP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LGALS3BP in serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LGALS3BP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LGALS3BP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LGALS3BP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15046 Supplier BIOHIPPO Supplier No. BHE15211588
Mouse LGALS3BP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LGALS3BP in serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LGALS3BP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LGALS3BP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LGALS3BP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color