Learn More
Sino Biological Anti-LIF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50755RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LIF (Catalog#50755-M08H; P09056; Met1-Phe203). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1637