Learn More
Sino Biological Anti-LIF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50756T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LIF (Catalog#50756-MNAH; NP_032527.1; Met1-Phe203). LIF specific IgG was purified by Mouse LIF affinity chromatography.
Catalog No. 50-241-0776